Learn More
Trajan Scientific SYRINGE 10UL HP GT RN 23/26
SDP
Catalog No. 502530381
Encompass_Preferred
SGE Autosampler Syringes with 42mm Cone Tip Removable Needle, Description: Gas tight, Volume: 10 uL, Diameter Outer Needle: 0.63/0.47 mm, Needle Gauge: 23 to 26 G, Length Needle: 42 mm, Connection Type: Removable, Diameter Outer: 0.63/0.47 mm
Catalog No. 50-253-0381 Supplier Trajan Scientific Supplier No. 002829
SGE Autosampler Syringes with 42mm Cone Tip Removable Needle, Description: Gas tight, Volume: 10 uL, Diameter Outer Needle: 0.63/0.47 mm, Needle Gauge: 23 to 26 G, Length Needle: 42 mm, Connection Type: Removable, Diameter Outer: 0.63/0.47 mm