Learn More
Trajan Scientific SYRINGE 1BR-11.5 1ML
SDP
Catalog No. 502530425
Encompass_Preferred
SGE NanoVolume Syringes, Volume: 1 uL, Needle Gauge: 23 G, Model: 1BR-11.5, Needle Point Style: Removable Cone, Length Needle: 115 mm, 4.52 in., Accuracy: +/-2%, Diameter Outer: 0.315 in., 8 mm, Diameter Outer Needle: 0.0248 in., 0.63 mm, Material: Borosilicate Glass, Quantity: Each
Catalog No. 50-253-0425 Supplier Trajan Scientific Supplier No. 000510
SGE NanoVolume Syringes, Volume: 1 uL, Needle Gauge: 23 G, Model: 1BR-11.5, Needle Point Style: Removable Cone, Length Needle: 115 mm, 4.52 in., Accuracy: +/-2%, Diameter Outer: 0.315 in., 8 mm, Diameter Outer Needle: 0.0248 in., 0.63 mm, Material: Borosilicate Glass, Quantity: Each