Learn More
SAI Infusion Technologies 1cc Syringe with attached 23G0.5" Blunt Needle - Sterile, 100/pack
SDP
Catalog No. NC9878077
Encompass
This sterile 1cc syringe comes with SAI's 23G x 0.5 inch blunt needle already attached, making it easy to maintain sterility while accessing catheters. Ideal for long-term infusion studies requiring flushing and locking of catheters. 100/pk. Product is for laboratory animal research only.
Catalog No. NC9878077 Supplier SAI Infusion Technologies Supplier No. B23501CC 100
This sterile 1cc syringe comes with SAI's 23G x 0.5 inch blunt needle already attached, making it easy to maintain sterility while accessing catheters. Ideal for long-term infusion studies requiring flushing and locking of catheters. 100/pk. Product is for laboratory animal research only.