Learn More
Med Vet International Terumo Syringe & Needle, 1mL, Removable Needle, 27G X 1/2in, Tuberculin, 100/BX, SS-01T2713
Catalog No. 502522596
Encompass_Preferred
Terumo SS-01T2713 1mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. The syringe contains a removable needle. This Terumo SS-01T2713 1mL Syringe & Needle Combo comes with a 27G X 1/2' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This is a tuberculin syringe. They are small syringes with fine needles used to administer medication under the skin and perform tuberculosis tests. This quantity is 100/BX
Catalog No. 50-252-2596 Supplier Med Vet International Supplier No. 01T2713
Terumo SS-01T2713 1mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. The syringe contains a removable needle. This Terumo SS-01T2713 1mL Syringe & Needle Combo comes with a 27G X 1/2' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This is a tuberculin syringe. They are small syringes with fine needles used to administer medication under the skin and perform tuberculosis tests. This quantity is 100/BX