Learn More
Med Vet International Easy Touch U-100 Insulin Syringe, 1mL 30g x 5/16in, 100/Box
Catalog No. NC1531943
Encompass_Preferred
The Easy Touch U-100 Insulin syringe is designed and manufactured to deliver the highest level of comfort. Their special needles are triple-bevel cut to a super sharp point, electro-polished to remove microscopic burrs, and film-coated for smoother injection. The attached 30g 5/16in. needle is color-coded to the gauge and the thin wall cannulas provide an easier flow. Color code: Blue Syringe size: 1cc/1mL Needle gauge: 30g Needle size: 5/16in.
This item is not returnable. View return policy
Catalog No. NC1531943 Supplier Med Vet International Supplier No. 830165
The Easy Touch U-100 Insulin syringe is designed and manufactured to deliver the highest level of comfort. Their special needles are triple-bevel cut to a super sharp point, electro-polished to remove microscopic burrs, and film-coated for smoother injection. The attached 30g 5/16in. needle is color-coded to the gauge and the thin wall cannulas provide an easier flow. Color code: Blue Syringe size: 1cc/1mL Needle gauge: 30g Needle size: 5/16in.