Learn More
Hach Company Syringe, luer slip tip, 1 cc
Supplier:  Hach Company 1226300
Encompass
Disposable syringes feature a luer slip tip suited for connection with syringe filters or filter holders. Marked graduations. 1/pk
Catalog No. 50-199-4869