Learn More
Trajan Scientific SYRINGE 1MF-LL-GT
SDP
Catalog No. 502550768
Encompass_Preferred
SGE Gastight Syringes: Fixed Luer-Lok Models, Volume: 1.0 mL, Diameter Outer: 0.31 in., 8.8 mm, Model: 1MDF-LL-GT, Needle Gauge: No Needle Included, Accuracy: +/-1%, Connection Type: Fixed, For Use With: Suitable for gases or liquids for maximum inertness
Catalog No. 50-255-0768 Supplier Trajan Scientific Supplier No. 008025
SGE Gastight Syringes: Fixed Luer-Lok Models, Volume: 1.0 mL, Diameter Outer: 0.31 in., 8.8 mm, Model: 1MDF-LL-GT, Needle Gauge: No Needle Included, Accuracy: +/-1%, Connection Type: Fixed, For Use With: Suitable for gases or liquids for maximum inertness