Learn More
Trajan Scientific SYRINGE 1MR-LC-GT
SDP
Catalog No. 502530308
Encompass_Preferred
SGE Manual Syringes For Rheodyne and Valco Valves, Connection Type: Removable, Volume: 1 mL, Model: 1MDR-LC-GT, Length Needle: 50.8 mm, 2 in., Needle Gauge: 22 G, Diameter Outer Needle: 0.028 in., 0.71 mm, For Use With: Rheodyne and Valco injection valves, Length: 2 in., Needle Point Style: LC
Catalog No. 50-253-0308 Supplier Trajan Scientific Supplier No. 008105
SGE Manual Syringes For Rheodyne and Valco Valves, Connection Type: Removable, Volume: 1 mL, Model: 1MDR-LC-GT, Length Needle: 50.8 mm, 2 in., Needle Gauge: 22 G, Diameter Outer Needle: 0.028 in., 0.71 mm, For Use With: Rheodyne and Valco injection valves, Length: 2 in., Needle Point Style: LC