Learn More
Trajan Scientific SYRINGE 1MR-V-GT
SDP
Catalog No. 502530307
Encompass_Preferred
SGE Syringes with Valves, Volume: 1 mL, Borosilicate Glass, PTFE Plunger, Length Needle: 50 mm, 2 in., Needle Gauge: 23 G, Connection Type: Removable, For Use With: For sample storage and transportation or to pressurize sample prior to injection, Length: 2 in., 50 mm, Needle Point Style: Cone
Catalog No. 50-253-0307 Supplier Trajan Scientific Supplier No. 008110
SGE Syringes with Valves, Volume: 1 mL, Borosilicate Glass, PTFE Plunger, Length Needle: 50 mm, 2 in., Needle Gauge: 23 G, Connection Type: Removable, For Use With: For sample storage and transportation or to pressurize sample prior to injection, Length: 2 in., 50 mm, Needle Point Style: Cone