Learn More
Trajan Scientific SYRINGE 2.5MF-LL-GT
SDP
Catalog No. 502530305
Encompass_Preferred
SGE Gastight Syringes: Fixed Luer-Lok Models, Volume: 2.5 mL, Diameter Outer: 0.42 in., 10.8 mm, Model: 2.5MDF-LL-GT, Needle Gauge: No Needle Included, Accuracy: +/-1%, Connection Type: Fixed, For Use With: Suitable for gases or liquids for maximum inertness
Catalog No. 50-253-0305 Supplier Trajan Scientific Supplier No. 008425
SGE Gastight Syringes: Fixed Luer-Lok Models, Volume: 2.5 mL, Diameter Outer: 0.42 in., 10.8 mm, Model: 2.5MDF-LL-GT, Needle Gauge: No Needle Included, Accuracy: +/-1%, Connection Type: Fixed, For Use With: Suitable for gases or liquids for maximum inertness