Learn More
Charter Medical SYRINGE 2 LONG PINCH CLP 96/CS
Catalog No. NC0475892
Encompass
Manufactured by Incell Corporation and supplied by Charter Medical:   2 in. long with pinch clamp, piercing pin and female luer lock (syringe) adapter
Catalog No. NC0475892 Supplier Charter Medical Supplier No. 0322092
Manufactured by Incell Corporation and supplied by Charter Medical:   2 in. long with pinch clamp, piercing pin and female luer lock (syringe) adapter