Learn More
Med Vet International Nipro Syringe, 20mL, Luer Lock, 50/Box, Jd+20L
Catalog No. 502832438
Encompass_Preferred
Nipro JD+20L 20mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 20mL syringe fits conventional and safety needles. This quantity is 50/BX
Catalog No. 50-283-2438 Supplier Med Vet International Supplier No. JD20L
Nipro JD+20L 20mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 20mL syringe fits conventional and safety needles. This quantity is 50/BX