Learn More
Med Vet International BD Syringe, 20mL, Luer Slip, 48/BX, 302831
Catalog No. 503044993
Encompass_Preferred
BD 302831 20mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 20mL Syringe fits conventional and safety needles. This quantity is 48/BX
Catalog No. 50-304-4993 Supplier Med Vet International Supplier No. 302831
BD 302831 20mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 20mL Syringe fits conventional and safety needles. This quantity is 48/BX