Learn More
Waters Corp Waters Syringe, 25 μL, HPLC System, Autosampler
Catalog No. NC9319407 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This syringe is an optional component for various autosamplers and separations modules, suitable for use with high-performance liquid chromatography (HPLC) systems.
  • Optional syringe for various autosamplers and separations modules
  • Designed for HPLC systems
  • Volume of 25 μL
  • Replacement as required
  • Compatible with 712, 715, 717, and 717 Plus autosamplers
  • Compatible with 2690, 2690D, 2695, 2695D, 2790, 2795 separations modules, and LC Module 1
This item is not returnable. View return policy
Catalog No. NC9319407 Supplier Waters Corp Supplier No. WAT077343
This syringe is an optional component for various autosamplers and separations modules, suitable for use with high-performance liquid chromatography (HPLC) systems.
  • Optional syringe for various autosamplers and separations modules
  • Designed for HPLC systems
  • Volume of 25 μL
  • Replacement as required
  • Compatible with 712, 715, 717, and 717 Plus autosamplers
  • Compatible with 2690, 2690D, 2695, 2695D, 2790, 2795 separations modules, and LC Module 1