Learn More
Trajan Scientific SYRINGE 250F 250UL FIXED
SDP
Catalog No. 502530323
Encompass_Preferred
SGE Series II Standard Fixed Needle Syringes, Volume: 250 uL, Needle Gauge: 25 G, Includes: Standard plunger, Length Needle: 50 mm, 1.96 in., Accuracy: +/-1%, Connection Type: Fixed, Diameter Outer: 0.31 in., 8 mm, Diameter Outer Needle: 0.01 in., 0.5 mm, Graduations: Major - 25.0 uL, Minor - 5.0 uL
Catalog No. 50-253-0323 Supplier Trajan Scientific Supplier No. 006000
SGE Series II Standard Fixed Needle Syringes, Volume: 250 uL, Needle Gauge: 25 G, Includes: Standard plunger, Length Needle: 50 mm, 1.96 in., Accuracy: +/-1%, Connection Type: Fixed, Diameter Outer: 0.31 in., 8 mm, Diameter Outer Needle: 0.01 in., 0.5 mm, Graduations: Major - 25.0 uL, Minor - 5.0 uL