Learn More
Trajan Scientific L-MARK Brand 250l, Collect PAL RN, NA, PTFE, Pt. Not
Catalog No. 50635363
Encompass_Preferred
L-MARK Brand Syringe, 250uL, For Collect PAL, 0.6 mm ID Plunger, PTFE, For up to 1mL/min applications; 10-32 Fittings at inlet and outlet; (see LMK.2620828 or 2620727 For Low Flow applications) Made in Germany.
Catalog No. 50-635-363 Supplier Trajan Scientific Supplier No. LMK.2620827
L-MARK Brand Syringe, 250uL, For Collect PAL, 0.6 mm ID Plunger, PTFE, For up to 1mL/min applications; 10-32 Fittings at inlet and outlet; (see LMK.2620828 or 2620727 For Low Flow applications) Made in Germany.