Learn More
Trajan Scientific SYRINGE 25F FN 25ML FIXED
SDP
Catalog No. 502530353
Encompass_Preferred
SGE Series II Standard Fixed Needle Syringes, Volume: 25 uL, Needle Gauge: 25 G, Includes: Standard plunger, Length Needle: 50 mm, 1.96 in., Accuracy: +/-1%, Connection Type: Fixed, Diameter Outer: 0.31 in., 8 mm, Diameter Outer Needle: 0.01 in., 0.5 mm, Graduations: Major - 2.5 uL, Minor - 0.5 uL
Catalog No. 50-253-0353 Supplier Trajan Scientific Supplier No. 003000
SGE Series II Standard Fixed Needle Syringes, Volume: 25 uL, Needle Gauge: 25 G, Includes: Standard plunger, Length Needle: 50 mm, 1.96 in., Accuracy: +/-1%, Connection Type: Fixed, Diameter Outer: 0.31 in., 8 mm, Diameter Outer Needle: 0.01 in., 0.5 mm, Graduations: Major - 2.5 uL, Minor - 0.5 uL