Learn More
Trajan Scientific SYRINGE 25F-GT
SDP
Catalog No. 502530351
Encompass_Preferred
SGE Gastight Syringes: Fixed Needle Models, Volume: 25 uL, Needle Gauge: 25 G, Model: 25F-GT, Length Needle: 50 mm, 1.96 in., Accuracy: +/-1%, Connection Type: Fixed, Diameter Outer: 0.31 in., 8 mm, Diameter Outer Needle: 0.01 in., 0.47 mm
Catalog No. 50-253-0351 Supplier Trajan Scientific Supplier No. 003200
SGE Gastight Syringes: Fixed Needle Models, Volume: 25 uL, Needle Gauge: 25 G, Model: 25F-GT, Length Needle: 50 mm, 1.96 in., Accuracy: +/-1%, Connection Type: Fixed, Diameter Outer: 0.31 in., 8 mm, Diameter Outer Needle: 0.01 in., 0.47 mm