Learn More
Thermo Scientific Syringes for UltiMate™ 3000 Autosamplers and Column Compartments
Catalog No. 501348314 Shop All Thermo Scientific Products
Encompass
Syringes for UltiMate™ 3000 Autosamplers and Column Compartments, 100μL capacity, the Thermo Scientific™ Dionex™ Syringes for UltiMate™ 3000 Autosamplers and Column Compartments are for use with WPS-3000 autosamplers and ACC-3000 column compartments.
Catalog No. 50-134-8314 Supplier Thermo Scientific Supplier No. 6822.0002
Syringes for UltiMate™ 3000 Autosamplers and Column Compartments, 100μL capacity, the Thermo Scientific™ Dionex™ Syringes for UltiMate™ 3000 Autosamplers and Column Compartments are for use with WPS-3000 autosamplers and ACC-3000 column compartments.