Learn More
Med Vet International AHS Syringe, 30mL, Luer Lock, 300/Case, AH30L
Catalog No. 503043202
Encompass_Preferred
AHS AH30L 30mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 30mL syringe fits conventional and safety needles. This quantity is 300/CS
Catalog No. 50-304-3202 Supplier Med Vet International Supplier No. AH30LX10
AHS AH30L 30mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 30mL syringe fits conventional and safety needles. This quantity is 300/CS