Learn More
Med Vet International BD Syringe, 30mL, Luer Slip, 56/BX, 302833
Catalog No. 503044522
Encompass_Preferred
BD 302833 30mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 30mL Syringe fits conventional and safety needles. This quantity is 56/BX
Catalog No. 50-304-4522 Supplier Med Vet International Supplier No. 302833
BD 302833 30mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 30mL Syringe fits conventional and safety needles. This quantity is 56/BX