Learn More
Med Vet International AHS Syringe, 30mL, Luer Slip, 300/Case, AH30S
Catalog No. 503045112
Encompass_Preferred
AHS AH30S 30mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 30mL syringe fits conventional and safety needles. This quantity is 300/CS
Catalog No. 50-304-5112 Supplier Med Vet International Supplier No. AH30SX10
AHS AH30S 30mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 30mL syringe fits conventional and safety needles. This quantity is 300/CS