Learn More
Med Vet International AHS Syringe, 30mL, Luer Slip, 30/Box, AH30S
Catalog No. 502522463
Encompass_Preferred
AHS AH30S 30mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 30mL syringe fits conventional and safety needles. This quantity is 30/BX
Catalog No. 50-252-2463 Supplier Med Vet International Supplier No. AH30S
AHS AH30S 30mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 30mL syringe fits conventional and safety needles. This quantity is 30/BX