Learn More
Med Vet International BD Syringe & Needle, 3mL, Luer Lock, 18G X 1 1/2", Hypodermic, 100/BX, 309580
Supplier:  Med Vet International 309580
Encompass_Preferred
BD 309580 3mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 3mL Syringe & Needle Combo fits conventional and safety needles. This BD 309580 3mL Syringe & Needle Combo comes with a 18G X 1 1/2" Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This contains a hypodermic needle which is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-304-4509