Learn More
Med Vet International BD Syringe & Needle, 3mL, Luer Lock, 23G X 1in, Hypodermic, 100/BX, 309571
Catalog No. 502522548
Encompass_Preferred
BD 309571 3mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 3mL Syringe & Needle Combo fits conventional and safety needles. This BD 309571 3mL Syringe & Needle Combo comes with a 23G X 1' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This contains a hypodermic needle which is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-252-2548 Supplier Med Vet International Supplier No. 309571
BD 309571 3mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 3mL Syringe & Needle Combo fits conventional and safety needles. This BD 309571 3mL Syringe & Needle Combo comes with a 23G X 1' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This contains a hypodermic needle which is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX