Learn More
Med Vet International Nipro Syringe & Needle, 3mL, Luer Lock, 25G X 1in, Hypodermic, 100/BX, JD+03L2525
Catalog No. 502522340
Encompass_Preferred
Nipro JD+03L2525 3mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 3mL syringe fits conventional and safety needles. This Nipro JD+03L2525 Syringe & Needle Combo comes with a 25G X 1' Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles allow rapid and efficient drug delivery. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-252-2340 Supplier Med Vet International Supplier No. JD03L2525
Nipro JD+03L2525 3mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 3mL syringe fits conventional and safety needles. This Nipro JD+03L2525 Syringe & Needle Combo comes with a 25G X 1' Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles allow rapid and efficient drug delivery. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX