Learn More
Med Vet International AHS Syringe & Needle, 3mL, Luer Lock, 25G X 5/8in, Hypodermic, 100/BX, AH03L2516
Supplier:  Med Vet International AH03L2516
Encompass_Preferred
AHS AH03L2516 3mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 3mL syringe fits conventional and safety needles. This AHS Needle is a 25G X 5/8' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-252-2473