Learn More
Med Vet International AHS Syringe & Needle, 3mL, Luer Lock, 25G X 1", Hypodermic, 1000/Case, AH03L2525
Catalog No. 503045176
Encompass_Preferred
AHS AH03L2525 3mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 3mL syringe fits conventional and safety needles. This AHS Needle is a 25G X 1" Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 1000/CS
Catalog No. 50-304-5176 Supplier Med Vet International Supplier No. AH03L2525X10
AHS AH03L2525 3mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 3mL syringe fits conventional and safety needles. This AHS Needle is a 25G X 1" Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 1000/CS