Learn More
Med Vet International Nipro Syringe, 3mL, Luer Lock, 100/Box, JD+03L
Catalog No. 502522343
Encompass_Preferred
Nipro JD+03L 3mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 3mL syringe fits conventional and safety needles. This quantity is 100/BX
Catalog No. 50-252-2343 Supplier Med Vet International Supplier No. JD03L
Nipro JD+03L 3mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 3mL syringe fits conventional and safety needles. This quantity is 100/BX