Learn More
Med Vet International 3mL (3Cc) Luer Lock Syringe With Safety Needle, 21g X 1in, 50/Box
Catalog No. 502830072
Encompass_Preferred
3mL (3cc) Luer Lock Syringe With Safety Needle, 21G X 1in, 50/Box Safety Needle Features One handed activation No solid surface required Simple finger stroke activation Protective arm keeps fingers behind the needle An audible "click" as well as tactile feel and a visual indicator confirm safety mechanism engagement Needle remains locked inside activated cover Fits any brand Syringe
Catalog No. 50-283-0072 Supplier Med Vet International Supplier No. 27105
3mL (3cc) Luer Lock Syringe With Safety Needle, 21G X 1in, 50/Box Safety Needle Features One handed activation No solid surface required Simple finger stroke activation Protective arm keeps fingers behind the needle An audible "click" as well as tactile feel and a visual indicator confirm safety mechanism engagement Needle remains locked inside activated cover Fits any brand Syringe