Learn More
Waters Corp Waters Quality Parts Syringe, 500 μL, for 2707 Autosampler
Catalog No. 50792463 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This syringe is designed for precise sample injections in the 2707 Autosampler.
  • Designed for precise sample injections.
  • Features a metal collar at both ends for ruggedness.
  • Steel plunger is less likely to break if bumped or dropped.
  • Compatible with 2707 Autosampler.
  • 500 μL volume.
  • Suitable for HPLC systems.
  • Replacement is as required.
Catalog No. 50-792-463 Supplier Waters Corp Supplier No. 700000862
This syringe is designed for precise sample injections in the 2707 Autosampler.
  • Designed for precise sample injections.
  • Features a metal collar at both ends for ruggedness.
  • Steel plunger is less likely to break if bumped or dropped.
  • Compatible with 2707 Autosampler.
  • 500 μL volume.
  • Suitable for HPLC systems.
  • Replacement is as required.