Learn More
Trajan Scientific SYRINGE 500R-LC
SDP
Catalog No. 502530312
Encompass_Preferred
SGE Manual Syringes For Rheodyne and Valco Valves, Connection Type: Removable, Volume: 500 uL, Model: 500R-LC, Length Needle: 50.8 mm, 2 in., Needle Gauge: 22 G, Diameter Outer Needle: 0.028 in., 0.71 mm, For Use With: Rheodyne and Valco injection valves, Length: 2 in., Needle Point Style: LC
Catalog No. 50-253-0312 Supplier Trajan Scientific Supplier No. 007310
SGE Manual Syringes For Rheodyne and Valco Valves, Connection Type: Removable, Volume: 500 uL, Model: 500R-LC, Length Needle: 50.8 mm, 2 in., Needle Gauge: 22 G, Diameter Outer Needle: 0.028 in., 0.71 mm, For Use With: Rheodyne and Valco injection valves, Length: 2 in., Needle Point Style: LC