Learn More
Trajan Scientific SYRINGE 50UL 50F-PE-0.63 AU
SDP
Catalog No. 502530338
Encompass_Preferred
SGE Thermo Scientific GC Autosampler Syringes, Connection Type: Fixed, Diameter Outer Needle: 0.024 in., 0.63 mm, Needle Gauge: 23 G, Length Needle: 2.7 in., 70 mm, Volume: 50 uL, For Use With: Gas chromatography, PerkinElmer, Length: 70 mm, Needle Point Style: Cone, Quantity: Each
Catalog No. 50-253-0338 Supplier Trajan Scientific Supplier No. 004670
SGE Thermo Scientific GC Autosampler Syringes, Connection Type: Fixed, Diameter Outer Needle: 0.024 in., 0.63 mm, Needle Gauge: 23 G, Length Needle: 2.7 in., 70 mm, Volume: 50 uL, For Use With: Gas chromatography, PerkinElmer, Length: 70 mm, Needle Point Style: Cone, Quantity: Each