Learn More
Trajan Scientific SYRINGE 5BR-5-RA8 5ML
SDP
Catalog No. 502530421
Encompass_Preferred
Trajan 5uL Plunger-in-Needle Syringes, Length Needle: 50 mm, Needle Gauge: 23 G, Volume: 5 uL, Diameter Outer Needle: 0.63 mm, Needle Point Style: Cone
Catalog No. 50-253-0421 Supplier Trajan Scientific Supplier No. 000850
Trajan 5uL Plunger-in-Needle Syringes, Length Needle: 50 mm, Needle Gauge: 23 G, Volume: 5 uL, Diameter Outer Needle: 0.63 mm, Needle Point Style: Cone