Learn More
Med Vet International AHS Syringe, 5mL, Luer Slip, 1000/Case, AH05S
Catalog No. 503045082
Encompass_Preferred
AHS AH05S 5mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 5mL syringe fits conventional and safety needles. This quantity is 1000/CS
Catalog No. 50-304-5082 Supplier Med Vet International Supplier No. AH05SX10
AHS AH05S 5mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 5mL syringe fits conventional and safety needles. This quantity is 1000/CS