Learn More
Med Vet International BD Syringe, 5mL, Luer Slip, 125/BX, 309647
Catalog No. 503043529
Encompass_Preferred
BD 309647 5mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 5mL Syringe fits conventional and safety needles. This quantity is 125/BX
Catalog No. 50-304-3529 Supplier Med Vet International Supplier No. 309647
BD 309647 5mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 5mL Syringe fits conventional and safety needles. This quantity is 125/BX