Learn More
Vici Precision Sampling, Inc. SYRINGE 5ML GAS
Catalog No. NC9843454
Encompass_Preferred
Gas Syringe A-2 5ml RN .029 in. x .012 in. x 2 in. bevel, open end
Catalog No. NC9843454 Supplier Vici Precision Sampling, Inc. Supplier No. 050035
Gas Syringe A-2 5ml RN .029 in. x .012 in. x 2 in. bevel, open end