Learn More
Med Vet International Dynarex #6989 Syringe Without Needle, 5Cc, Luer Lock, 100/Box
Catalog No. 502830447
Encompass_Preferred
Syringe Without Needle, Luer Lock (5mL/cc) - item #6989, This syringe offers a clear barrel with bold scale marking for ease of measurement. This item is blister packed to ensure sterility, non-toxic, pyrogen and latex free. ? Available in a wide range of gauges? Clear barrel w/bold scale marking? Blister packed for sterility? Non-toxic, pyrogen & latex free
Catalog No. 50-283-0447 Supplier Med Vet International Supplier No. 6989
Syringe Without Needle, Luer Lock (5mL/cc) - item #6989, This syringe offers a clear barrel with bold scale marking for ease of measurement. This item is blister packed to ensure sterility, non-toxic, pyrogen and latex free. ? Available in a wide range of gauges? Clear barrel w/bold scale marking? Blister packed for sterility? Non-toxic, pyrogen & latex free