Learn More
Chemglass Life Sciences Syringe, Polypropylene, 5mL Capacity, Centric Tip
SDP
Catalog No. CG308004
Encompass_Preferred
Syringe, Polypropylene, 5mL Capacity, Centric Tip. These all-polypropylene syringes are latex free and contain no rubber or silicone. The plastic plunger has a smooth draw and a positive safety stop to prevent accidental spills. Syringes are individually sterile strip packed. The Luer tip for the 5mL sizes is centric to the barrel.
Catalog No. CG308004 Supplier Chemglass Life Sciences Supplier No. CG308004
Syringe, Polypropylene, 5mL Capacity, Centric Tip. These all-polypropylene syringes are latex free and contain no rubber or silicone. The plastic plunger has a smooth draw and a positive safety stop to prevent accidental spills. Syringes are individually sterile strip packed. The Luer tip for the 5mL sizes is centric to the barrel.