Learn More
Trajan Scientific SYRINGE 5MR-GT
SDP
Catalog No. 502550767
Encompass_Preferred
SGE Gastight Syringes: Removable-Needle Models, Diameter Outer: 0.53 in., 13.7 mm, Needle Gauge: 23 G, Volume: 5.0 mL, Needle Point Style: Removable Bevel, Length Needle: 50 mm, 1.96 in., Accuracy: +/-1%, Connection Type: Removable, Diameter Outer Needle: 0.02 in., 0.63 mm
Catalog No. 50-255-0767 Supplier Trajan Scientific Supplier No. 008700
SGE Gastight Syringes: Removable-Needle Models, Diameter Outer: 0.53 in., 13.7 mm, Needle Gauge: 23 G, Volume: 5.0 mL, Needle Point Style: Removable Bevel, Length Needle: 50 mm, 1.96 in., Accuracy: +/-1%, Connection Type: Removable, Diameter Outer Needle: 0.02 in., 0.63 mm