Learn More
Trajan Scientific SYRINGE 5MR-VLLMA-GT
SDP
Catalog No. 502530298
Encompass_Preferred
SGE Syringes with Valves, Volume: 5 mL, Borosilicate Glass, PTFE Plunger, Autoclavable: Autoclavable, Connection Type: Removable, For Use With: For sample storage and transportation or to pressurize sample prior to injection, Termination: Luer-lock
Catalog No. 50-253-0298 Supplier Trajan Scientific Supplier No. 008770
SGE Syringes with Valves, Volume: 5 mL, Borosilicate Glass, PTFE Plunger, Autoclavable: Autoclavable, Connection Type: Removable, For Use With: For sample storage and transportation or to pressurize sample prior to injection, Termination: Luer-lock