Learn More
Trajan Scientific L-MARK Brand 5l Syringe For Agilent, FN, 23g, Pt. AS, 6 Pack
Catalog No. 501179815
Encompass_Preferred
L-MARK Brand Syringe, For Agilent 7673, 7683, 7693A, 6850 AutoSamplers, Similar to Agilent Part Number 5182-0875, 5uL, FN, 23s, 42mm, Pt. AS, Fitted Plunger, 6 Pack Made in Germany.
Catalog No. 50-117-9815 Supplier Trajan Scientific Supplier No. LMK.3030133
L-MARK Brand Syringe, For Agilent 7673, 7683, 7693A, 6850 AutoSamplers, Similar to Agilent Part Number 5182-0875, 5uL, FN, 23s, 42mm, Pt. AS, Fitted Plunger, 6 Pack Made in Germany.