Learn More
Trajan Scientific SYRINGE 5UL .63MMOD 5F-HP-0.63
SDP
Supplier:  Trajan Scientific 001810
Encompass_Preferred
SGE Autosampler Syringes with 42 mm Cone Tip Fixed Needle, 5 uL, Description: Not gas tight, Diameter Outer Needle: 0.63 mm, Needle Gauge: 23 G, Connection Type: Fixed, For Use With: Agilent GC Autosamplers HP7673, 7683, 7693A & 6850ALS, Gas chromatography, Length Needle: 42 mm, Volume: 5 uL
Catalog No. 50-253-0417