Learn More
Trajan Scientific L-MARK Brand 5l Syringe For Agilent, FN, 23s-26s, 42mm, Pt. AS
Catalog No. 501179810
Encompass_Preferred
L-MARK Brand Syringe, For Agilent 7673, 7683, 7693A, 6850 AutoSamplers, Similar to Agilent Part Number 5181-1273, 5uL, Fixed Needle (FN), 23s-26s, 42mm, Point Style AS, Fitted Plunger, 3 Pack Made in Germany.
Catalog No. 50-117-9810 Supplier Trajan Scientific Supplier No. LMK.3030127
L-MARK Brand Syringe, For Agilent 7673, 7683, 7693A, 6850 AutoSamplers, Similar to Agilent Part Number 5181-1273, 5uL, Fixed Needle (FN), 23s-26s, 42mm, Point Style AS, Fitted Plunger, 3 Pack Made in Germany.