Learn More
Med Vet International AHS Syringe, 60mL, Catheter, 25/Box, AH60C
Catalog No. NC1402885
Encompass_Preferred
AHS AH60C 60mL Syringe comes with a clear barrel and easy to read graduation marks. The catheter tip is used for flushing catheters, gastostomy tubes and other devices. This quantity is 25/BX
Catalog No. NC1402885 Supplier Med Vet International Supplier No. AH60C
AHS AH60C 60mL Syringe comes with a clear barrel and easy to read graduation marks. The catheter tip is used for flushing catheters, gastostomy tubes and other devices. This quantity is 25/BX