Learn More
Med Vet International Syringe, 60 mL, l/l 40/each
Catalog No. 502643623
Encompass_Preferred
Luer Lock Syringe BarrelsFits All Luer Lock Needles• Luer Lock Hub Provides a Secure Connectionto Prevent Unintentional Disengagement andAvoid Leaks• Easy-to-Read Barrel Markings for AccurateMedication Dosage
This item is not returnable. View return policy
Catalog No. 50-264-3623 Supplier Med Vet International Supplier No. 802064
Luer Lock Syringe BarrelsFits All Luer Lock Needles• Luer Lock Hub Provides a Secure Connectionto Prevent Unintentional Disengagement andAvoid Leaks• Easy-to-Read Barrel Markings for AccurateMedication Dosage