Learn More
Perkin Elmer US LLC AutoSystem Autosampler Syringe, Metal Plunger in Barrel with PTFE-Tipped Seal
Catalog No. 509045122
Encompass_Preferred
These autosampler syringes are for routine analyses. The metal plunger in barrel with PTFE-tipped seal is the standard syringe as shipped with each Clarus GC instrument. Recommended use: Routine analyses. Shipped standard with instrument. Plunger type: metal plunger barrel with Tetlon-tipped seal. Required for complete installation.
Catalog No. 50-904-5122 Supplier Perkin Elmer US LLC Supplier No. N6101390
These autosampler syringes are for routine analyses. The metal plunger in barrel with PTFE-tipped seal is the standard syringe as shipped with each Clarus GC instrument. Recommended use: Routine analyses. Shipped standard with instrument. Plunger type: metal plunger barrel with Tetlon-tipped seal. Required for complete installation.