Learn More
Perkin Elmer US LLC AutoSystem Autosampler Syringe, Plunger Type: Metal Plunger in Barrel, Volume: 5µL, Outer Diameter: 0.63
Catalog No. 509045108
Encompass_Preferred
Recommended autosampler syringes are available in 0.5, 5 and 50 µL capacities. For routine analyses, the metal plunger in barrel with PTFE-tipped seal is the standard syringe as shipped with each Clarus GC instrument. Alternative syringes to use are the metal plunger in barrel or the 0.53 mm on-column injection.
Catalog No. 50-904-5108 Supplier Perkin Elmer US LLC Supplier No. N6101251
Recommended autosampler syringes are available in 0.5, 5 and 50 µL capacities. For routine analyses, the metal plunger in barrel with PTFE-tipped seal is the standard syringe as shipped with each Clarus GC instrument. Alternative syringes to use are the metal plunger in barrel or the 0.53 mm on-column injection.