Learn More
Thermo Scientific Syringe Adapter Kit 1 Pk
Catalog No. NC9621723 Shop All Thermo Scientific Products
Encompass
syringe adapter kit contains plastic adapters in different sizes and configurations that fit on syringe tip and have ports on opposite end compatible with standard medical fittings to interface syringes with IV lines, catheters, feeding tubes, or other clinical apparatus for injecting fluids or medications, made of polypropylene for single-use.
This item is not returnable. View return policy
Catalog No. NC9621723 Supplier Thermo Scientific Supplier No. 7000562011
syringe adapter kit contains plastic adapters in different sizes and configurations that fit on syringe tip and have ports on opposite end compatible with standard medical fittings to interface syringes with IV lines, catheters, feeding tubes, or other clinical apparatus for injecting fluids or medications, made of polypropylene for single-use.