Learn More
B BRAUN MED INC OEM OnGuard 2 Syringe Adaptor, needle-free Luer Lock syringe adapter, white winged, 16G fluid path, 0.041 mL dead volume, latex-free, 50 per box (300 per case)
Catalog No. NC2182668
Encompass_Preferred
A needle-free syringe adaptor designed as a secure, leak-proof IV connector for safe fluid and medication administration that helps reduce accidental needlestick injuries. It uses self-sealing elastomers for dry disconnection to prevent drug leakage and provides an efficient fluid pathway for vial and IV transfers.
  • Secure, leak-proof connection with an audible click on engagement.
  • Luer lock compatible for broad attachment to syringes and IV sets.
  • 16 gauge fluid pathway for efficient withdrawal and transfer of liquids.
  • Dead volume 0.041 ml to minimize residual fluid.
  • Winged design with protective cap for safer handling and bolus delivery.
  • Latex-free construction suitable for those with latex sensitivities.
  • Compatible with 20 mm and 13 mm vials.
  • Packaging: 50 units per box, 300 units per case; shelf life 36 months.
  • This item is not returnable. View return policy
    Catalog No. NC2182668 Supplier B BRAUN MED INC OEM Supplier No. 412163
    A needle-free syringe adaptor designed as a secure, leak-proof IV connector for safe fluid and medication administration that helps reduce accidental needlestick injuries. It uses self-sealing elastomers for dry disconnection to prevent drug leakage and provides an efficient fluid pathway for vial and IV transfers.
  • Secure, leak-proof connection with an audible click on engagement.
  • Luer lock compatible for broad attachment to syringes and IV sets.
  • 16 gauge fluid pathway for efficient withdrawal and transfer of liquids.
  • Dead volume 0.041 ml to minimize residual fluid.
  • Winged design with protective cap for safer handling and bolus delivery.
  • Latex-free construction suitable for those with latex sensitivities.
  • Compatible with 20 mm and 13 mm vials.
  • Packaging: 50 units per box, 300 units per case; shelf life 36 months.
  • This item is not returnable. View return policy